missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-benzyloxycarbonyl-D-Tryptophan, ≥98 %, Thermo Scientific™

Catalog Number p-7044930
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2279-15-4
C19H18N2O4
338.36
MFCD00037945
AHYFYYVVAXRMKB-QGZVFWFLSA-N
z-d-trp-oh, n-cbz-d-tryptophan, z-d-tryptophan, cbz-d-trp-oh, nalpha-cbz-d-tryptophan, r-2-benzyloxy carbonyl amino-3-1h-indol-3-yl propanoic acid, n a-benzyloxycarbonyl-d-tryptophan, chembl65134, nalpha-carbobenzoxy-d-tryptophan, d-tryptophan, n-phenylmethoxy carbonyl
700663
(2R)-3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propansäure
OC(=O)[C@@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2279-15-4
C19H18N2O4
338.36
MFCD00037945
AHYFYYVVAXRMKB-QGZVFWFLSA-N
z-d-trp-oh, n-cbz-d-tryptophan, z-d-tryptophan, cbz-d-trp-oh, nalpha-cbz-d-tryptophan, r-2-benzyloxy carbonyl amino-3-1h-indol-3-yl propanoic acid, n a-benzyloxycarbonyl-d-tryptophan, chembl65134, nalpha-carbobenzoxy-d-tryptophan, d-tryptophan, n-phenylmethoxy carbonyl
700663
(2R)-3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propansäure
OC(=O)[C@@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N(alpha)-Benzyloxycarbonyl-D-tryptophan
Schmelzpunkt 122°C to 124°C
Beilstein 96744
Formelmasse 338.37
Reinheit (%) ≥98%
Menge 250 mg
Optische Drehung −3.1° (c=3 in Acetic Acid)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'