missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-beta-Alanin-Ethylester, 95 %, Thermo Scientific™

Catalog Number p-7047117
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
23583-21-3
C12H17NO2
207.273
MFCD00085541
HCTJHQFFNDLDPF-UHFFFAOYSA-N
ethyl 3-benzylamino propanoate, ethyl 3-benzylamino propionate, ethyl n-benzyl-beta-alaninate, n-benzyl-beta-alanine ethyl ester, 3-benzylamino-propionic acid ethyl ester, 3-benzylaminopropionic acid ethyl ester, n-benzyl-3-aminopropionic acid ethyl ester, ethyl n-benzyl-, a-alaninate, ethyl3-benzylamino propanoate
90196
Ethyl 3-(benzylamino)propanoat
CCOC(=O)CCNCC1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
23583-21-3
C12H17NO2
207.273
MFCD00085541
HCTJHQFFNDLDPF-UHFFFAOYSA-N
ethyl 3-benzylamino propanoate, ethyl 3-benzylamino propionate, ethyl n-benzyl-beta-alaninate, n-benzyl-beta-alanine ethyl ester, 3-benzylamino-propionic acid ethyl ester, 3-benzylaminopropionic acid ethyl ester, n-benzyl-3-aminopropionic acid ethyl ester, ethyl n-benzyl-, a-alaninate, ethyl3-benzylamino propanoate
90196
Ethyl 3-(benzylamino)propanoat
CCOC(=O)CCNCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Benzyl-beta-alanine ethyl ester
Siedepunkt 150°C (0.2mmHg)
Strukturformel C6H5CH2NHCH2CH2CO2CH2CH3
Brechungsindex 1.505
Formelmasse 207.27
Reinheit (%) 98%
Dichte 1.03
Menge 1 g
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'