missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-6-Aminohexanosäure, 99 %, Thermo Scientific Chemicals

Catalog Number p-7044987
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1947-00-8
C14H19NO4
265.309
MFCD00004423
RXQDBVWDABAAHL-UHFFFAOYSA-N
z-6-aminohexanoic acid, z-epsilon-acp-oh, 6-benzyloxy carbonyl amino hexanoic acid, n-benzyloxycarbonyl-6-aminocaproic acid, cbz-, a-aminocaproic acid, n-cbz-6-aminohexanoic acid, n-benzyloxycarbonyl-6-aminohexanoic acid, n-carbobenzoxy-epsilon-aminocaproic acid, 6-phenylmethoxy carbonyl amino hexanoic acid
74758
6-(Phenylmethoxycarbonylamino)hexansäure
C1=CC=C(C=C1)COC(=O)NCCCCCC(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1947-00-8
C14H19NO4
265.309
MFCD00004423
RXQDBVWDABAAHL-UHFFFAOYSA-N
z-6-aminohexanoic acid, z-epsilon-acp-oh, 6-benzyloxy carbonyl amino hexanoic acid, n-benzyloxycarbonyl-6-aminocaproic acid, cbz-, a-aminocaproic acid, n-cbz-6-aminohexanoic acid, n-benzyloxycarbonyl-6-aminohexanoic acid, n-carbobenzoxy-epsilon-aminocaproic acid, 6-phenylmethoxy carbonyl amino hexanoic acid
74758
6-(Phenylmethoxycarbonylamino)hexansäure
C1=CC=C(C=C1)COC(=O)NCCCCCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Benzyloxycarbonyl-6-aminohexanoic acid
Schmelzpunkt 53°C to 57°C
Menge 5 g
Beilstein 2288508
Formelmasse 265.31
Reinheit (%) 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'