missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-Alanin-Methyl-Ester, 95 %, Thermo Scientific™

Catalog Number p-7041594
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
28819-05-8
C12H15NO4
237.255
MFCD00270464
OMDVFTVXPVXANK-VIFPVBQESA-N
z-ala-ome, z-l-alanine methyl ester, methyl 2s-2-benzyloxy carbonyl amino propanoate, n-trityl-l-ala, cbz-ala-ome, z-l-alaninemethylester, n-z-alanine methyl ester, n-cbz-l-alanine methyl ester, methyl n-benzyloxycarbonylalaninate, n-carbobenzyloxy-l-alanine methyl ester
6993447
Methyl-(2S)-2-(phenylmethoxycarbonylamino)propanoat
CC(C(=O)OC)NC(=O)OCC1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
28819-05-8
C12H15NO4
237.255
MFCD00270464
OMDVFTVXPVXANK-VIFPVBQESA-N
z-ala-ome, z-l-alanine methyl ester, methyl 2s-2-benzyloxy carbonyl amino propanoate, n-trityl-l-ala, cbz-ala-ome, z-l-alaninemethylester, n-z-alanine methyl ester, n-cbz-l-alanine methyl ester, methyl n-benzyloxycarbonylalaninate, n-carbobenzyloxy-l-alanine methyl ester
6993447
Methyl-(2S)-2-(phenylmethoxycarbonylamino)propanoat
CC(C(=O)OC)NC(=O)OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Benzyloxycarbonyl-L-alanine methyl ester
Menge 5 g
Formelmasse 237.25
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'