missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1-Boc-L-tryptophan, 95%

Product Code. 15469058
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15469058 1 g 1g
15479058 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15469058

Brand: Thermo Scientific Alfa Aesar H62542.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 144599-95-1 | C21H28N2O6 | 404.46 g/mol

N-Boc-1-Boc-L-tryptophan is generally used as a intermediate in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1-Boc-L-tryptophan is generally used as a intermediate in pharmaceutical and chemical research.

Solubility
Soluble in dimethylformamide (1 mmole in 2 ml).

Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144599-95-1
Molecular Formula C21H28N2O6
Molecular Weight (g/mol) 404.46
MDL Number MFCD00270520
InChI Key FATGZMFSCKUQGO-HNNXBMFYSA-N
Synonym boc-trp boc-oh, s-3-1-tert-butoxycarbonyl-1h-indol-3-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-l-trp boc-oh, ambotzbaa1140, nalpha,1-di-boc-l-tryptophan, n,n'-di-t-butyloxycarbonyl-l-tryptophane, 2s-2-2-methylpropan-2-yl oxycarbonylamino-3-1-2-methylpropan-2-yl oxycarbonyl indol-3-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-tert-butoxycarbonyl indol-3-yl propanoic acid, 2s-3-1-tert-butoxy carbonyl-1h-indol-3-yl-2-tert-butoxy carbonyl amino propanoic acid
PubChem CID 7020330
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CN(C(=O)OC(C)(C)C)C2=CC=CC=C12)C(O)=O

Specifications

Quantity 1 g
Solubility Information Soluble in dimethylformamide (1 mmole in 2 ml).
Formula Weight 404.46
Percent Purity 95%
Chemical Name or Material N-Boc-1-Boc-L-tryptophan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.