missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1-(p-toluolsulfonyl)-L-Histidin, 98 %, Thermo Scientific™

Catalog Number 15436178
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
35899-43-5
C18H23N3O6S
409.46
MFCD00065967
DCLJSEPKYJSEHW-UHFFFAOYNA-N
boc-his tos-oh, boc-l-his tos-oh, boc-his tos, boc-l-histidine tosyl, s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid, boc-his-tos, ambotzbaa1153, pubchem7363, boc-his-tos-oh
6455010
(2S)-3-[1-(4-Methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propansäure
CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(CC(NC(=O)OC(C)(C)C)C(O)=O)=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
35899-43-5
C18H23N3O6S
409.46
MFCD00065967
DCLJSEPKYJSEHW-UHFFFAOYNA-N
boc-his tos-oh, boc-l-his tos-oh, boc-his tos, boc-l-histidine tosyl, s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid, boc-his-tos, ambotzbaa1153, pubchem7363, boc-his-tos-oh
6455010
(2S)-3-[1-(4-Methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propansäure
CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(CC(NC(=O)OC(C)(C)C)C(O)=O)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Boc-1-(p-toluenesulfonyl)-L-histidine
Schmelzpunkt ∽125°C (decomposition)
Menge 1 g
Beilstein 769957
Optische Drehung +16° (c=1 in Methanol)
Formelmasse 409.46
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'