missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-Fluoro-L-Phenylalanin, 98 %, Thermo Scientific Chemicals

Catalog Number p-7041570
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
114873-00-6
C14H18FNO4
283.299
MFCD00672520
NTWUXBKUDXGMHV-NSHDSACASA-N
boc-phe 2-f-oh, boc-l-2-fluorophenylalanine, boc-2-fluoro-l-phenylalanine, boc-l-2-fluorophe, boc-l-phe 2-f-oh, s-n-boc-2-fluorophenylalanine, boc-d-2-fluorophenylalanine, s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid, boc-2-fluoro-l-phe, 2s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid
2734496
(2S)-3-(2-Fluorphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propansäure
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1F)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
114873-00-6
C14H18FNO4
283.299
MFCD00672520
NTWUXBKUDXGMHV-NSHDSACASA-N
boc-phe 2-f-oh, boc-l-2-fluorophenylalanine, boc-2-fluoro-l-phenylalanine, boc-l-2-fluorophe, boc-l-phe 2-f-oh, s-n-boc-2-fluorophenylalanine, boc-d-2-fluorophenylalanine, s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid, boc-2-fluoro-l-phe, 2s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid
2734496
(2S)-3-(2-Fluorphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propansäure
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1F)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Boc-2-fluoro-L-phenylalanine
Schmelzpunkt 92°C to 96°C
Menge 1 g
Beilstein 5347990
Optische Drehung +9° (c=1 in Ethyl Acetate)
Formelmasse 283.3
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'