missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2 -Methyl-D-Serin, 97 %, Thermo Scientific Chemicals

Catalog Number p-7035082
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
84311-18-2
C9H17NO5
219.237
MFCD02682596
FWRXDSRYWWYTPD-SECBINFHSA-N
r-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoic acid, n-boc-alpha-methyl-d-serine, boc-alpha-methyl-d-ser, boc-me-d-ser-oh, 2r-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoic acid, boc-d-alpha-methylserine, boc-a-methyl-d-ser, n-boc-a-methyl-d-serine, n-boc-2-methyl-d-serine, n-tert-butoxycarbonyl-2-methyl-d-serine
24749404
(2R)-3-Hydroxy-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propansäure
CC(C)(C)OC(=O)NC(C)(CO)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
84311-18-2
C9H17NO5
219.237
MFCD02682596
FWRXDSRYWWYTPD-SECBINFHSA-N
r-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoic acid, n-boc-alpha-methyl-d-serine, boc-alpha-methyl-d-ser, boc-me-d-ser-oh, 2r-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoic acid, boc-d-alpha-methylserine, boc-a-methyl-d-ser, n-boc-a-methyl-d-serine, n-boc-2-methyl-d-serine, n-tert-butoxycarbonyl-2-methyl-d-serine
24749404
(2R)-3-Hydroxy-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propansäure
CC(C)(C)OC(=O)NC(C)(CO)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Boc-2-methyl-D-serine
Menge 250 mg
Formelmasse 219.24
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'