missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-Beta-Alanin, 99%, Thermo Scientific Chemicals

Catalog Number p-7026499
Click to view available options
Menge:
2.5 g
10 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3303-84-2
C8H15NO4
189.21
MFCD00037291
WCFJUSRQHZPVKY-UHFFFAOYSA-N
boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid
76809
3-[(2-Methylpropan-2-yl)oxycarbonylamino]propansäure
CC(C)(C)OC(=O)NCCC(O)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3303-84-2
C8H15NO4
189.21
MFCD00037291
WCFJUSRQHZPVKY-UHFFFAOYSA-N
boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid
76809
3-[(2-Methylpropan-2-yl)oxycarbonylamino]propansäure
CC(C)(C)OC(=O)NCCC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Boc-beta-alanine
Schmelzpunkt 73°C to 76°C
Strukturformel (CH3)3CO2CNHCH2CH2CO2H
Formelmasse 189.21
Reinheit (%) 99%
Menge 2.5 g

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'