missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-DL-Alanin, 97 %, Thermo Scientific Chemicals

Catalog Number 15488808
Click to view available options
Menge:
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3744-87-4
C8H15NO4
189.211
MFCD00063124
QVHJQCGUWFKTSE-UHFFFAOYSA-N
boc-dl-alanine, boc-dl-ala-oh, 2-tert-butoxycarbonyl amino propanoic acid, n-boc-dl-alanine, 2-n-boc-amino-propionic acid, n-tert-butoxycarbonyl alanine, 2-tert-butoxycarbonylamino-propionic acid, 2-tert-butoxycarbonylamino propanoic acid, 2-tert-butoxy carbonyl amino propanoic acid
268471
2-[(2-Methylpropan-2-yl)oxycarbonylamino]propansäure
CC(C(=O)O)NC(=O)OC(C)(C)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3744-87-4
C8H15NO4
189.211
MFCD00063124
QVHJQCGUWFKTSE-UHFFFAOYSA-N
boc-dl-alanine, boc-dl-ala-oh, 2-tert-butoxycarbonyl amino propanoic acid, n-boc-dl-alanine, 2-n-boc-amino-propionic acid, n-tert-butoxycarbonyl alanine, 2-tert-butoxycarbonylamino-propionic acid, 2-tert-butoxycarbonylamino propanoic acid, 2-tert-butoxy carbonyl amino propanoic acid
268471
2-[(2-Methylpropan-2-yl)oxycarbonylamino]propansäure
CC(C(=O)O)NC(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Boc-DL-alanine
Menge 100 g
Formelmasse 189.21
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'