missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-(S)-Cyclopropylalanin Benzylester, 95 %

Catalog Number 15487525
Click to view available options
Menge:
250 mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Boc-(S)-cyclopropylalanine benzyl ester
CAS 406681-37-6
Schmelzpunkt 52°C to 54°C
Summenformel C18H25NO4
MDL-Nummer MFCD08060934
Menge 250 mg
Synonym n-boc-s-cyclopropylalanine benzyl ester, s-benzyl 2-tert-butoxycarbonyl cyclopropyl amino propanoate, benzyl 2s-2-tert-butoxycarbonyl cyclopropyl amino propanoate, cyclopropanepropanoicacid, a-1,1-dimethylethoxy carbonyl amino-,phenylmethyl ester, as
InChI-Schlüssel KRQPCZKCAZIODC-ZDUSSCGKSA-N
SMILES C[C@H](N(C1CC1)C(=O)OC(C)(C)C)C(=O)OCC1=CC=CC=C1
IUPAC-Name Benzyl-(2S)-2-[cyclopropyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoat
Molekulargewicht (g/mol) 319.40
PubChem CID 21134475
Formelmasse 319.4
Reinheit (%) 95%
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'