missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-Trans-4-Hydroxy-D-Prolin, 99 %, Thermo Scientific Chemicals

Catalog Number p-7029664
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
147266-92-0
C10H17NO5
231.25
MFCD01861341
BENKAPCDIOILGV-UHFFFAOYNA-N
2r,4s-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, n-boc-trans-4-hydroxy-d-proline, boc-trans-4-hydroxy-d-proline, boc-d-hyp-oh, unii-7k1v4sdl4l, 7k1v4sdl4l, 2r,4s-1-tert-butoxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 4s-1-tert-butoxycarbonyl-4-hydroxy-d-proline, n-boc-4-hydroxy-d-proline, trans, 2r,4s-n-alpha-t-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid
688615
(2R,4S)-4-Hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidin-2-carbonsäure
CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
147266-92-0
C10H17NO5
231.25
MFCD01861341
BENKAPCDIOILGV-UHFFFAOYNA-N
2r,4s-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, n-boc-trans-4-hydroxy-d-proline, boc-trans-4-hydroxy-d-proline, boc-d-hyp-oh, unii-7k1v4sdl4l, 7k1v4sdl4l, 2r,4s-1-tert-butoxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 4s-1-tert-butoxycarbonyl-4-hydroxy-d-proline, n-boc-4-hydroxy-d-proline, trans, 2r,4s-n-alpha-t-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid
688615
(2R,4S)-4-Hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidin-2-carbonsäure
CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Boc-trans-4-hydroxy-D-proline
Schmelzpunkt ∼123°C
Menge 250 mg
Formelmasse 231.25
Reinheit (%) 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'