missing translation for 'onlineSavingsMsg'
Learn More

N-Dodecyl-acetamide, TRC

Product Code. 30378296
Change view
Click to view available options
Menge:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30378296 10 mg
30370888 100 mg
30370551 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378296 Supplier Toronto Research Chemicals Supplier No. TRCD56205010MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3886-80-4
Summenformel C14H29NO
Molekulargewicht (g/mol) 227.39
SMILES CCCCCCCCCCCCNC(=O)C

Specifications

Chemischer Name oder Material N-Dodecyl-acetamide
Menge 10 mg
InChI-Formel InChI=1S/C14H29NO/c1-3-4-5-6-7-8-9-10-11-12-13-15-14(2)16/h3-13H2,1-2H3,(H,15,16)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.