missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-Methionin, 98 %, Thermo Scientific™

Catalog Number p-7041459
Click to view available options
Menge:
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
112883-40-6
C20H21NO4S
371.45
MFCD00062958 MFCD00037134 MFCD00062958
BUBGAUHBELNDEW-GOSISDBHSA-N
fmoc-d-met-oh, fmoc-d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl-d-methionine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylthio butanoic acid, d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, n-fmoc-d-methionine, ambotzfaa1145, n-9-fluorenylmethyloxycarbonyl-d-methionine
6992521
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfanylbutansäure
CSCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
112883-40-6
C20H21NO4S
371.45
MFCD00062958 MFCD00037134 MFCD00062958
BUBGAUHBELNDEW-GOSISDBHSA-N
fmoc-d-met-oh, fmoc-d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl-d-methionine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylthio butanoic acid, d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, n-fmoc-d-methionine, ambotzfaa1145, n-9-fluorenylmethyloxycarbonyl-d-methionine
6992521
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfanylbutansäure
CSCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Fmoc-D-methionine
Schmelzpunkt 130°C to 134°C
Formelmasse 371.45
Reinheit (%) 98%
Menge 5 g
Optische Drehung +30° (c=1 in DMF)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'