missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1-Naphthalinmethylaminhydrochlorid, 98 %, Thermo Scientific Chemicals

Catalog Number p-7030436
Click to view available options
Menge:
1 g
10 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
65473-13-4
C12H14ClN
207.701
MFCD00012555
BVJVHPKFDIYQOU-UHFFFAOYSA-N
n-methyl-1-naphthalenemethylamine hydrochloride, n-methyl-1-naphthalen-1-yl methanamine hydrochloride, n-methyl-1-naphthylmethylamine hydrochloride, n-methyl-n-naphthylmethylamine hydrochloride, unii-z8vrt9b309, methyl naphthalen-1-ylmethyl amine hydrochloride, n-methyl-1-naphthalenemethanamine hydrochloride, n-methyl-c-naphthalen-1-yl methanamine hydrochloride, 1-naphthalenemethanamine, n-methyl-, hydrochloride 1:1, 1-methylaminomethyl naphthalene hydrochloride
16211748
N-methyl-1-naphthalen-1-ylmethanamin;Hydrochlorid
CNCC1=CC=CC2=CC=CC=C21.Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
65473-13-4
C12H14ClN
207.701
MFCD00012555
BVJVHPKFDIYQOU-UHFFFAOYSA-N
n-methyl-1-naphthalenemethylamine hydrochloride, n-methyl-1-naphthalen-1-yl methanamine hydrochloride, n-methyl-1-naphthylmethylamine hydrochloride, n-methyl-n-naphthylmethylamine hydrochloride, unii-z8vrt9b309, methyl naphthalen-1-ylmethyl amine hydrochloride, n-methyl-1-naphthalenemethanamine hydrochloride, n-methyl-c-naphthalen-1-yl methanamine hydrochloride, 1-naphthalenemethanamine, n-methyl-, hydrochloride 1:1, 1-methylaminomethyl naphthalene hydrochloride
16211748
N-methyl-1-naphthalen-1-ylmethanamin;Hydrochlorid
CNCC1=CC=CC2=CC=CC=C21.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Methyl-1-naphthalenemethylamine hydrochloride
Schmelzpunkt 190°C to 193°C
Menge 1 g
Formelmasse 207.7
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'