missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-(1-piperazinyl)-nicotinamid, 99 %, Thermo Scientific™

Catalog Number p-7033798
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
902837-15-4
C11H16N4O
220.28
MFCD08061094
PKJNZUNKHVUUBC-UHFFFAOYSA-N
n-methyl-2-piperazin-1-yl-nicotinamide, n-methyl-2-1-piperazinyl nicotinamide, n-methyl-2-piperazin-1-yl pyridine-3-carboxamide, n-methyl-2-1-piperazino nicotinamide, n-methyl-2-piperazin-1-yl nicotinamide, n-methyl 2-piperazinyl 3-pyridyl carboxamide, n-methyl-2-piperazin-1-ylnicotinamide, n-methyl-2-1-piperazinyl pyridine-3-carboxamide
28054516
N-Methyl-2-(piperazin-1-yl)pyridin-3-carboxamid
CNC(=O)C1=C(N=CC=C1)N1CCNCC1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
902837-15-4
C11H16N4O
220.28
MFCD08061094
PKJNZUNKHVUUBC-UHFFFAOYSA-N
n-methyl-2-piperazin-1-yl-nicotinamide, n-methyl-2-1-piperazinyl nicotinamide, n-methyl-2-piperazin-1-yl pyridine-3-carboxamide, n-methyl-2-1-piperazino nicotinamide, n-methyl-2-piperazin-1-yl nicotinamide, n-methyl 2-piperazinyl 3-pyridyl carboxamide, n-methyl-2-piperazin-1-ylnicotinamide, n-methyl-2-1-piperazinyl pyridine-3-carboxamide
28054516
N-Methyl-2-(piperazin-1-yl)pyridin-3-carboxamid
CNC(=O)C1=C(N=CC=C1)N1CCNCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Methyl-2-(1-piperazinyl)nicotinamide
Menge 250 mg
Formelmasse 220.28
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'