missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-2-(1-piperazinyl)-nicotinamid Hydrochlorid, 96 %, Thermo Scientific™

Catalog Number p-7034032
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
902836-08-2
C12H19ClN4O
270.761
MFCD12407084
YRPLJKUUYNCGQS-UHFFFAOYSA-N
n,n-dimethyl-2-piperazin-1-yl nicotinamide hydrochloride, n,n-dimethyl-2-1-piperazinyl nicotinamide hydrochloride, n,n-dimethyl-2-piperazin-1-yl pyridine-3-carboxamide hydrochloride, n,n-dimethyl-2-1-piperazinyl nicotinamide hcl, n,n-dimethyl-2-1-piperazinyl pyridine-3-carboxamide hydrochloride, n,n-dimethyl-2-piperazin-1-yl-pyridine-3-carboxamide hydrochloride, n,n-dimethyl-2-piperazin-1-yl pyridine-3-carboxamide-hydrogen chloride 1/1
53418086
N,N-Dimethyl-2-Piperazin-1-ylpyridin-3-Carboxamid;Hydrochlorid
CN(C)C(=O)C1=C(N=CC=C1)N2CCNCC2.Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
902836-08-2
C12H19ClN4O
270.761
MFCD12407084
YRPLJKUUYNCGQS-UHFFFAOYSA-N
n,n-dimethyl-2-piperazin-1-yl nicotinamide hydrochloride, n,n-dimethyl-2-1-piperazinyl nicotinamide hydrochloride, n,n-dimethyl-2-piperazin-1-yl pyridine-3-carboxamide hydrochloride, n,n-dimethyl-2-1-piperazinyl nicotinamide hcl, n,n-dimethyl-2-1-piperazinyl pyridine-3-carboxamide hydrochloride, n,n-dimethyl-2-piperazin-1-yl-pyridine-3-carboxamide hydrochloride, n,n-dimethyl-2-piperazin-1-yl pyridine-3-carboxamide-hydrogen chloride 1/1
53418086
N,N-Dimethyl-2-Piperazin-1-ylpyridin-3-Carboxamid;Hydrochlorid
CN(C)C(=O)C1=C(N=CC=C1)N2CCNCC2.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N,N-Dimethyl-2-(1-piperazinyl)nicotinamide hydrochloride
Menge 250 mg
Formelmasse 270.76
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'