Learn More
N-Nitrosopiperidine, TRC
Product Code. 30041314
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041314 500 mg 500mg
30040662 1 g 1g
30005984 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30041314 Supplier Toronto Research Chemicals Supplier No. TRCN545370500MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100-75-4
Molecular Formula C5 H10 N2 O
Molecular Weight (g/mol) 114.15
Synonym Piperidine, 1-nitroso-, 1-Nitrosopiperidine, N-Nitrosopiperidine, NPI, NPIP, NSC 138
IUPAC Name 1-nitrosopiperidine
SMILES O=NN1CCCCC1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C5H10N2O/c8-6-7-4-2-1-3-5-7/h1-5H2
Formula Weight 114.0793
Percent Purity >95%
Chemical Name or Material N-Nitrosopiperidine
Grade HPLC