missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAB2 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 17722354
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17722354 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17722354 Lieferant Invitrogen™ Lieferanten-Nr. PA5120954

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Positive test controls include: M21, Mouse lung, Mouse liver. The target is usually found in the following locations: Nucleus. Immunogen sequence: MHRAPSPTAE QPPGGGDSAR RTLQPRLKPS ARAMALPRTL GELQLYRVLQ RANLLSYYET FIQQGGDDVQ QLCEAGEEEF LEIMALVGMA TKPLHVRRLQ KALREWATNP GLFSQPVPAV PVSSIPLFKI SETAGTRKGS MSNGHGSPGE KAGSARSFSP KSPLELGEKL SPLPGGPGAG DPRIWPGRST PESDVGAGGE.

Transcriptional control is in part regulated by interactions between DNA-bound transcription factors, such as Egr-1/NGFI-A, and coregulatory proteins, such as NAB (for NGFI-A-binding proteins). The evolutionarily conserved NAB proteins, NAB1 and NAB2, are corepressors of EGF-1/NGFI-A. Both NAB1 and NAB2 contain an amino terminal NAB conserved domain 1 (NCB1), which is required for binding NGFI-A, and a carboxy terminal NCD2 domain, which is responsible for the repressor function of NAB proteins. NAB2 is principally localized in the nucleus and may play a role in the downregulation of NGFI-A activity as well as in controlling fundamental processes such as cell division, differentiation and apoptosis. NAB2 has a predicted molecular mass of 56 kDa and localizes to chromosome 12q13.3-14.1, a region that is rearranged in several solid tumors, lipomas and liposarcomas.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen NAB2
Anwendungen ELISA, Western Blot
Klassifikation Polyclonal
Konzentration 1.36 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen NAB2
Gen-Zugriffsnummer Q15742, Q61127
Gen-Alias AI451907; EGR1 binding protein 2; EGR-1-binding protein 2; MADER; Melanoma-associated delayed early response protein; NAB2; NGFI-A binding protein 2; NGFI-A binding protein 2 (EGR1 binding protein 2); NGFI-A binding protein 2 (EGR-1 binding protein 2); NGFI-A-binding protein 2; Protein MADER
Gensymbole NAB2
Wirtsspezies Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human NAB2 (NP_005958.1).
Reinigungsverfahren Affinity Chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 17937, 4665
Zielspezies Human, Mouse
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.