missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Ndelta-Trityl-L-Glutamin Pentafluorphenyl-Ester, 97 %, Thermo Scientific™

Catalog Number p-7029690
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
132388-65-9
C45H33F5N2O5
776.76
MFCD00800874
OLYKAAWLZTVISS-DHUJRADRSA-N
fmoc-gln trt-opfp, fmoc-l-gln trt-opfp, fmoc-glutamine trt-opfp, fmoc-n-delta-trityl-l-glutamine pentafluorophenyl ester, fmoc-n-gamma-trityl-l-glutamine pentafluorophenyl ester, n-fmoc-n'-trityl-l-glutamine pentafluorophenyl ester, n, a-fmoc-n, a-trityl-l-glutamine pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-tri, n-alpha-fmoc-gamma-trityl-l-glutamine pentafluorophenyl ester
11297250
(2,3,4,5,6-Pentafluorphenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoat
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)OC4=C(C(=C(C(=C4F)F)F)F)F)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
132388-65-9
C45H33F5N2O5
776.76
MFCD00800874
OLYKAAWLZTVISS-DHUJRADRSA-N
fmoc-gln trt-opfp, fmoc-l-gln trt-opfp, fmoc-glutamine trt-opfp, fmoc-n-delta-trityl-l-glutamine pentafluorophenyl ester, fmoc-n-gamma-trityl-l-glutamine pentafluorophenyl ester, n-fmoc-n'-trityl-l-glutamine pentafluorophenyl ester, n, a-fmoc-n, a-trityl-l-glutamine pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-tri, n-alpha-fmoc-gamma-trityl-l-glutamine pentafluorophenyl ester
11297250
(2,3,4,5,6-Pentafluorphenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoat
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)OC4=C(C(=C(C(=C4F)F)F)F)F)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Nalpha-Fmoc-Ndelta-trityl-L-glutamine pentafluorophenyl ester
Schmelzpunkt 183°C to 184°C
Menge 1 g
Optische Drehung −16° (c=1 in THF)
Formelmasse 776.75
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'