missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Ngamma-Trityl-L-Asparagin, 97 %, Thermo Scientific Chemicals

Catalog Number p-7039984
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
132388-59-1
C38H31N2O5
595.68
MFCD00077049
KJYAFJQCGPUXJY-UMSFTDKQSA-M
fmoc-asn trt-oh, nalpha-fmoc-ngamma-trityl-l-asparagine, ambotzfaa1015, n-fmoc-n4-trityl-l-asparagine, pubchem9947, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-oxo-4-tritylamino butanoic acid, fmoc-n'-trityl-l-asparagine, nalpha-9h-fluoren-9-ylmethoxy carbonyl-ngamma-trityl-l-asparagine, n-9-fluorenyl methoxy carbonyl-n-trityl-l-asparagine
640248
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[(triphenylmethyl)carbamoyl]propanoate
[O-]C(=O)[C@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
132388-59-1
C38H31N2O5
595.68
MFCD00077049
KJYAFJQCGPUXJY-UMSFTDKQSA-M
fmoc-asn trt-oh, nalpha-fmoc-ngamma-trityl-l-asparagine, ambotzfaa1015, n-fmoc-n4-trityl-l-asparagine, pubchem9947, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-oxo-4-tritylamino butanoic acid, fmoc-n'-trityl-l-asparagine, nalpha-9h-fluoren-9-ylmethoxy carbonyl-ngamma-trityl-l-asparagine, n-9-fluorenyl methoxy carbonyl-n-trityl-l-asparagine
640248
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[(triphenylmethyl)carbamoyl]propanoate
[O-]C(=O)[C@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Nalpha-Fmoc-Ngamma-trityl-L-asparagine
Schmelzpunkt 201°C to 204°C
Menge 5 g
Beilstein 4343823
Optische Drehung −15° (c=1 in Methanol)
Formelmasse 596.67
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'