missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPSA Mouse anti-Human, Clone: 1H7F2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 60259-2-IG-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Napsin 1 is an aspartic protease that is generally expressed in the lung, but also present in kidney and in metastatic carcinomas throughout the body. Napsin 1 processes surfactant protein B in healthy pulmonary and renal tissues, but is abnormally regulated when carcinogenesis occurs. Thus, many researchers have found napsin evaluation to be useful in identifying several cancers, and differentiating primary tumors from metastases. When combined with thyroid transcription factor analysis, napsin can be used as a specific biomarker for differentiation of carcinoma types. Napsin has also been associated with pulmonary alveolar proteinosis as well as in protein catabolism in renal tubules.Spezifikation
NAPSA | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O96009 | |
NAPSA | |
NAPSA Fusion Protein Ag9721 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin) | |
1H7F2 | |
Unconjugated | |
NAPSA | |
Asp 4, ASP4, Aspartyl protease 4, KAP, Kdap, NAP1, NAPA, NAPSA, Napsin 1, Napsin A, napsin A aspartic peptidase, Pronapsin A, SNAPA, TA01/TA02 | |
Mouse | |
Protein G | |
RUO | |
9476 | |
-20°C | |
Liquid |