missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAT5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15807-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
NAT5 is a component of N-acetyltransferase complex B (NatB). Human NatB performs cotranslational N(alpha)-terminal acetylation of methionine residues when they are followed by asparagine (Starheim et al., 2008Spezifikation
NAT5 | |
Polyclonal | |
Unconjugated | |
NAA20 | |
N acetyltransferase 5, NAA20, NAT3, NAT5, NatB complex subunit NAT5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362228, 51126, 67877 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61599, P61600 | |
NAA20 | |
NAT5 Fusion Protein Ag8541 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |