missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAM1/CD56 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E8C9, Proteintech
Mouse Monoclonal Antibody
140.00 € - 399.00 €
Spezifikation
Antigen | NCAM1/CD56 |
---|---|
Klon | 1E8C9 |
Konzentration | 0.95 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16887265
|
Proteintech
60238-1-IG-20UL |
20 μL |
140.00 €
20 Mikroliter |
Verfügbar ab: 29-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16877265
|
Proteintech
60238-1-IG-150UL |
150 μL |
399.00 €
150 Mikroliter |
Verfügbar ab: 29-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
NCAM, as a member of the immunoglobulin superfamily of adhesion molecules is characterized by several immunoglobulin (Ig)-like domains. The extracellular part of NCAM consists of five of these Ig domains and two fibronectin type III homology regions. NCAM is encoded by a single copy gene composed of 26 exons. However, at least 20-30 distinct isoforms can be generated by alternative splicing and by posttranslational modifications, such as sialylation. During sialylation, polysialic acid (PSA) carbohydrates are attached to the extracellular part of NCAM. Through its extracellular region, NCAM mediates homophilic interactions. In addition, NCAM can also undergo heterophilic interactions by binding extracellular matrix components, such as laminin, or other cell adhesion molecules, such as integrins.Spezifikation
NCAM1/CD56 | |
0.95 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD56, MSK39, N CAM 1, NCAM, NCAM 1, NCAM1 | |
Ncam1 | |
IgG1 | |
Protein G | |
Antibody |
1E8C9 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Porcine, Rat | |
P13591, P13595, P13596 | |
100515564, 17967, 24586, 4684 | |
NCAM1/CD56 Fusion Protein Ag5732 | |
Primary | |
-20°C | |
NCAM1 |