missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCOA6 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 15780021 Alle ansehen Thermo Scientific Produkte
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15780021 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15780021 Lieferant Invitrogen™ Lieferanten-Nr. PA552071

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Immunogen sequence: NQNVQHAGGQ GAGPPQNQMQ VSHGPPNMMQ PSLMGIHGNM NNQQAGTSGV PQVNLSNMQG QPQQGPPSQL MGMHQQIVPS QGQMVQQQGT LNPQNPMILS RAQLMPQGQM MVNPPSQNLG PSPQRMTPPK Q Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

Nuclear receptor coactivator that directly binds nuclear receptors and stimulates the transcriptional activities in a hormone-dependent fashion. Coactivates expression in an agonist- and AF2-dependent manner. Involved in the coactivation of different nuclear receptors, such as for steroids (GR and ERs), retinoids (RARs and RXRs), thyroid hormone (TRs), vitamin D3 (VDR) and prostanoids (PPARs). Probably functions as a general coactivator, rather than just a nuclear receptor coactivator. May also be involved in the coactivation of the NF-kappa-B pathway. May coactivate expression via a remodeling of chromatin and its interaction with histone acetyltransferase proteins.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen NCOA6
Anwendungen Immunohistochemistry (Paraffin), Immunocytochemistry
Klassifikation Polyclonal
Konzentration 0.1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gen NCOA6
Gen-Zugriffsnummer Q14686
Gen-Alias activating signal cointegrator 2; activating signal cointegrator-2; AIB3; Amplified in breast cancer protein 3; amplified in breast cancer-3 protein; ASC2; ASC-2; Cancer-amplified transcriptional coactivator ASC-2; KIAA0181; mKIAA0181; NCOA6; Ncoa7; NRC; NRC RAP250; nuclear receptor coactivator 6; Nuclear receptor coactivator RAP250; nuclear receptor-activating protein 250; nuclear receptor-activating protein, 250 kDa; peroxisome proliferator-activated receptor interacting protein; peroxisome proliferator-activated receptor interacting protein,PRIP; peroxisome proliferator-activated receptor-interacting protein; PPAR interacting protein PRIP; PPAR-interacting protein; PRIP; RAP250; thyroid hormone receptor binding protein; thyroid hormone receptor-binding protein; TRBP
Gensymbole NCOA6
Wirtsspezies Rabbit
Immunogen Recombinant protein corresponding to Human NCOA6. Recombinant protein control fragment (Product #RP-102323).
Reinigungsverfahren Antigen Affinity Chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 23054
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.