missing translation for 'onlineSavingsMsg'
Learn More

NDP Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 15380-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16894534

  • 140.00 € / 20 Mikroliter
Verfügbar ab: 04-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody detects 11-15 kDa (monomer) and 48 kDa (polymers).

Norrie disease is an X-linked genetic disorder characterized by progressive atrophy of the eyes, mental disturbances and deafness. The gene responsible for this disease was initially identified through positional cloning. Norrin, the gene product, encodes a small secreted, cysteine-rich protein that is thought to act as a ligand for the Wnt-receptor/beta-catenin signal pathway despite having sequence homology with the Wnt family of proteins. Mice lacking this gene have abnormal blood vessel growth in the vitreous and a disorganized retina, transgenic ectopic expression of Norrin restores normal retinal vasculature. Recent evidence shows that Norrin can attenuate tPA and uPA-mediated death of transformed rat retinal ganglion cells (RGC-5) by activating the Wnt/beta-catenin pathway and regulating the phosphorylation of LRP-1, a cell surface receptor for tPA and uPA, suggesting the Norrin may function in vivo by regulating kinases which may alter the phosphorylation of LRP-1.
Spezifikation

Spezifikation

NDP
Polyclonal
Unconjugated
NDP
EVR2, FEVR, ND, NDP, Norrie disease (pseudoglioma), Norrie disease protein, Norrin
Rabbit
Antigen Affinity Chromatography
RUO
17986, 4693
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P48744, Q00604
Ndp
NDP Fusion Protein Ag4066
20 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote