missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Spezifikation
Antigen | NDP |
---|---|
Konzentration | 0.3 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16894534
|
Proteintech
15380-1-AP-20UL |
20 μL |
140.00 €
20 Mikroliter |
Verfügbar ab: 23-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16884534
|
Proteintech
15380-1-AP-150UL |
150 μL |
399.00 €
150 Mikroliter |
Verfügbar ab: 23-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
This antibody detects 11-15 kDa (monomer) and 48 kDa (polymers).
Norrie disease is an X-linked genetic disorder characterized by progressive atrophy of the eyes, mental disturbances and deafness. The gene responsible for this disease was initially identified through positional cloning. Norrin, the gene product, encodes a small secreted, cysteine-rich protein that is thought to act as a ligand for the Wnt-receptor/beta-catenin signal pathway despite having sequence homology with the Wnt family of proteins. Mice lacking this gene have abnormal blood vessel growth in the vitreous and a disorganized retina, transgenic ectopic expression of Norrin restores normal retinal vasculature. Recent evidence shows that Norrin can attenuate tPA and uPA-mediated death of transformed rat retinal ganglion cells (RGC-5) by activating the Wnt/beta-catenin pathway and regulating the phosphorylation of LRP-1, a cell surface receptor for tPA and uPA, suggesting the Norrin may function in vivo by regulating kinases which may alter the phosphorylation of LRP-1.Spezifikation
NDP | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P48744, Q00604 | |
17986, 4693 | |
NDP Fusion Protein Ag4066 | |
Primary | |
-20°C | |
NDP |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
EVR2, FEVR, ND, NDP, Norrie disease (pseudoglioma), Norrie disease protein, Norrin | |
Ndp | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |