missing translation for 'onlineSavingsMsg'
Learn More

NDP Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 15380-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16884534

  • 329.85 € / 150 Mikroliter

Voraussichtliches Versanddatum: 23-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody detects 11-15 kDa (monomer) and 48 kDa (polymers).

Norrie disease is an X-linked genetic disorder characterized by progressive atrophy of the eyes, mental disturbances and deafness. The gene responsible for this disease was initially identified through positional cloning. Norrin, the gene product, encodes a small secreted, cysteine-rich protein that is thought to act as a ligand for the Wnt-receptor/beta-catenin signal pathway despite having sequence homology with the Wnt family of proteins. Mice lacking this gene have abnormal blood vessel growth in the vitreous and a disorganized retina, transgenic ectopic expression of Norrin restores normal retinal vasculature. Recent evidence shows that Norrin can attenuate tPA and uPA-mediated death of transformed rat retinal ganglion cells (RGC-5) by activating the Wnt/beta-catenin pathway and regulating the phosphorylation of LRP-1, a cell surface receptor for tPA and uPA, suggesting the Norrin may function in vivo by regulating kinases which may alter the phosphorylation of LRP-1.
Spezifikation

Spezifikation

NDP
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P48744, Q00604
EVR2, FEVR, ND, NDP, Norrie disease (pseudoglioma), Norrie disease protein, Norrin
Rabbit
IgG
150 μL
-20°C
Primary
17986, 4693
Western Blot
Unconjugated
NDP
Liquid
Ndp
NDP Fusion Protein Ag4066
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate