Learn More
Nectin-4/PVRL4 Antibody (337516), mFluor Violet 610 SE, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Nectin-4/PVRL4 Monoclonal antibody specifically detects Nectin-4/PVRL4 in Human samples. It is validated for Flow Cytometry, CyTOF
Spezifikation
Spezifikation
| Antigen | Nectin-4/PVRL4 |
| Anwendungen | Flow Cytometry, CyTOF |
| Klassifikation | Monoclonal |
| Klon | 337516 |
| Konjugat | mFluor Violet 610 SE |
| Verdünnung | Flow Cytometry, CyTOF-ready |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | EDSS1, Ig superfamily receptor LNIR, LNIRnectin 4, nectin-4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4DKFZp686K05193 |
| Wirtsspezies | Mouse |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant human Nectin-4, Gly27-Val351, Accession # Q96NY8 |
| Mehr anzeigen |