Beschreibung
Neurogranin Polyclonal specifically detects Neurogranin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spezifikation
Spezifikation
| Verdünnung | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Gensymbole | NRGN |
| Reinigungsverfahren | Affinity Purified |