missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamidoxim, 97 %, Thermo Scientific™

Catalog Number p-7034100
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1594-58-7
C6H7N3O
137.14
MFCD00265955
AQBMQGDKWIPBRF-UHFFFAOYSA-N
3-pyridylamidoxime, 3-pyridinecarboxamidoxime, z-n'-hydroxynicotinimidamide, n-hydroxynicotinimidamide, z-n'-hydroxypyridine-3-carboximidamide, n-hydroxy-nicotinamidine, 3-pyridinecarboxamidoxine, 3-pyridinecarboxamide oxime, nicotinamidoxime, hydroxyimino-3-pyridylmethylamine
5372334
(Z)-N'-hydroxypyridine-3-carboximidamide
N\C(=N/O)C1=CC=CN=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1594-58-7
C6H7N3O
137.14
MFCD00265955
AQBMQGDKWIPBRF-UHFFFAOYSA-N
3-pyridylamidoxime, 3-pyridinecarboxamidoxime, z-n'-hydroxynicotinimidamide, n-hydroxynicotinimidamide, z-n'-hydroxypyridine-3-carboximidamide, n-hydroxy-nicotinamidine, 3-pyridinecarboxamidoxine, 3-pyridinecarboxamide oxime, nicotinamidoxime, hydroxyimino-3-pyridylmethylamine
5372334
(Z)-N'-hydroxypyridine-3-carboximidamide
N\C(=N/O)C1=CC=CN=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Nicotinamidoxime
Schmelzpunkt 130°C to 135°C
Menge 1 g
Formelmasse 137.14
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'