Learn More
Niflumic Acid, TRC
Product Code. 30132596
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
5 g
Unit Size:
10mg
2.5mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132596 2.5 mg 2.5mg
30135460 5 g 5g
18042750 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132596 Supplier Toronto Research Chemicals Supplier No. TRCN4575002.5MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4394-00-7
Molecular Formula C13 H9 F3 N2 O2
Molecular Weight (g/mol) 282.22
Synonym Niflumic Acid, 2-[[3-(Trifluoromethyl)phenyl]amino]-3-pyridinecarboxylic acid, 2-(α,α,α-Trifluoro-m-toluidino)nicotinic acid, 2-[3-(Trifluoromethyl)anilino]nicotinic acid, UP-83, Actol, Forenol, Landruma, Nifluril
IUPAC Name 2-[3-(trifluoromethyl)anilino]pyridine-3-carboxylic acid
SMILES OC(=O)c1cccnc1Nc2cccc(c2)C(F)(F)F
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C13H9F3N2O2/c14-13(15,16)8-3-1-4-9(7-8)18-11-10(12(19)20)5-2-6-17-11/h1-7H,(H,17,18)(H,19,20)
Formula Weight 282.0616
Chemical Name or Material Niflumic Acid