missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16361-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The nucleolus is a dense subnuclear membraneless organelle that assembles around clusters of rRNA genes and functions in ribosome biogenesis. This gene encodes a nucleolar RNA-associated protein that is highly conserved between species. RNase treatment of permeabilized cells indicates that the nucleolar localization is RNA dependent. Further studies suggest that the protein is associated with ribosome biogenesis through an interaction with pre-rRNA primary transcripts. Alternative splicing has been observed at this locus and two splice variants encoding distinct isoforms have been identified.Spezifikation
NOL6 | |
Polyclonal | |
Unconjugated | |
NOL6 | |
NOL6, NRAP, Nucleolar protein 6, UTP22 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230082, 313167, 65083 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R5K4, Q9H6R4 | |
NOL6 | |
NOL6 Fusion Protein Ag8368 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |