missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NOP56 Polyclonal antibody specifically detects NOP56 in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | NOP56 |
| Anwendungen | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:500 - 1:2000, ELISA |
| Zusammensetzung | PBS (pH 7.3), 50% glycerol |
| Gen-Alias | NOL5A, NOP56 ribonucleoprotein homolog (yeast), nucleolar protein 56, Nucleolar protein 5A, nucleolar protein 5A (56kD with KKE/D repeat), nucleolar protein 5A (56kDa with KKE/D repeat) |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human NOP56 (NP_006383.2).,, Sequence:, MVLLHVLFEHAVGYALLALKEVEEISLLQPQVEESVLNLGKFHSIVRLVAFCPFASSQVALENANAVSEGVVHEDLRLLLETHLPSKKKKVLLGVGDPKIGAAIQEELGYNCQTGGVIAEILRGVRLHFHNLVKGLTDLSACKAQLGLGHSYSRAKVKFN |
| Reinigungsverfahren | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?