missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Nope/IGDCC4 Antibody, R&D Systems™
Rat Monoclonal Antibody
N.V.
Spezifikation
Antigen | Nope/IGDCC4 |
---|---|
Klon | 261123 |
Konzentration | LYOPH |
Verdünnung | Western Blot 1 ug/mL |
Anwendungen | Western Blot |
Beschreibung
Nope/IGDCC4 Monoclonal specifically detects Nope/IGDCC4 in Mouse samples. It is validated for Western Blot.Spezifikation
Nope/IGDCC4 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q9EQS9 | |
57722 | |
Mouse myeloma cell line NS0-derived recombinant mouse Nope/IGDCC4, Gly22-His953, Accession # Q9EQS9 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Nope/IGDCC4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse (rm) DCC, rmNeogenin, and rmPunc is observed. |
261123 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DDM36, hDDM36, IGDCC4, immunoglobulin superfamily DCC subclass member 4, immunoglobulin superfamily, DCC subclass, member 4, KIAA1628, likely ortholog of mouse neighbor of Punc E11, LOC57722, Neighbor of punc e11, NOPEFLJ42051, Protein DDM36 | |
IGDCC4 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only