Learn More
Invitrogen™ Novex™ EASIA™ MMP-3 Human ELISA Kit
Quantifies protein levels of human MMP-3 in serum, plasma, supernatant, and synovial fluid.
Marke: Invitrogen™ KAC1541
Weitere Details : Netto-Gewicht : 0.50000kg
Beinhaltet: Pre-coated plate, standard, controls, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Beschreibung
Matrix metalloproteinase-3 (MMP-3) degrades connective tissue matrix.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Bestellinformation
Shipping Condition: Room Temperature
Spezifikation
NM_002422.3, NP_002413.1 | |
Plasma, Serum, Supernatant, Synovial Fluid | |
Microplate Reader | |
3 hr. | |
MMP-3 | |
matrix metallopeptidase 3 |
<0.1ng/mL | |
Colorimetric Detection | |
4314 | |
Chromogen, Controls, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer | |
96 Assays |
For Research Use Only. Not for use in diagnostic procedures.