Learn More
Invitrogen™ Novex IL-17 Mouse ELISA Kit
Designed to quantify protein levels of mouse IL-17 in serum, plasma, and supernatant
Marke: Invitrogen™ KMC3021
Weitere Details : Netto-Gewicht : 0.47000kg
Beinhaltet: Pre-coated plate, standard, controls, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Beschreibung
Interleukin-17 (IL-17) plays an important role in bridging innate and adaptive immunity.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Bestellinformation
Shipping Condition: Wet ice
Spezifikation
Plasma, Serum, Supernatant | |
16171 | |
<5pg/mL | |
Interleukin 17A |
Colorimetric | |
Chromogen, Controls, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer | |
96 Assays | |
4 hr. |
For Research Use Only. Not for use in diagnostic procedures.