Learn More
Invitrogen™ Novex™ IL-23 Heterodimer Human ELISA Kit
Quantifies natural and recombinant human IL-23, but not the monomeric forms of the p19 or p40 subunits in serum, plasma, buffered solution, or cell culture medium
Marke: Invitrogen™ KHC0231
Weitere Details : Netto-Gewicht : 0.50000kg
Beinhaltet: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers
Beschreibung
IL-23 is important in the maturation of memory T-cells.
Kit is an alernate for Cat. No. KHC0232.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Bestellinformation
Shipping Condition: Room Temperature
Spezifikation
NM_002187.2, NP_002178.2 | |
Plasma, Serum, Supernatant | |
Microplate Reader | |
4 hr. | |
IL-23 | |
interleukin 12B |
<20pg/mL | |
Colorimetric Detection | |
3593 | |
Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer | |
96 Assays |
For Research Use Only. Not for use in diagnostic procedures.