Learn More
Invitrogen™ Novex IL-4 Monkey ELISA Kit
Key cytokine in the immune network
Marke: Invitrogen™ KPC0041
Weitere Details : Netto-Gewicht : 0.70000kg
Beinhaltet: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Beschreibung
The IL-4 Monkey ELISA Kit quantifies natural and recombinant monkey IL-4 in monkey serum, plasma, buffered solution, or cell culture medium.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Bestellinformation
Shipping Condition: Wet ice
Spezifikation
Plasma, Serum, Supernatant | |
3565 | |
<3pg/mL | |
For research use only (RUO) | |
Interleukin 4 |
Colorimetric | |
Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer | |
96 Assays | |
50 μL , 100 μL supernatant | |
4 hr. |
For Research Use Only. Not for use in diagnostic procedures.