Learn More
Invitrogen™ Luminex™ TNF- Human Singleplex Bead Kit
Designed for the measurement of human TNF-α in serum, plasma, and tissue culture supernatant
Marke:  Invitrogen™ LHC3011
Weitere Details : Netto-Gewicht : 0.04000kg
Artikelnummer. 10446433
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »
Beinhaltet: Capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet.
Benötigt : Buffer reagents needed to complete the reaction are sold separately under Catalog number LHB0001.
Beschreibung
Beschreibung
This assay may be run alone or in combination with other select Luminex™ singleplex kits from Life Technologies. The panel is suitable for use with the Luminex 100™/200™ and FLEXMAP 3D™ systems.
  • The TNF-α Human Singleplex Bead Kit is based on xMAP™ technology
  • The use of a suspension bead-based technology enables the multiplexing capabilities of the Luminex™ assays
  • Polystyrene microspheres are internally dyed with red and infrared fluorophores of differing intensities
  • Each bead is given a unique number, or bead region, allowing differentiation of one bead from another
  • Beads covalently bound to different antibodies can be mixed in the same assay, utilizing a 96-well microplate format
  • At the completion of the sandwich immunoassay, beads can be read using the detection systems
  • The TNF-α Human Singleplex Bead Kit has been validated using the same rigorous criterion that distinguishes Luminex assays from other commercially available assays
Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Immunoassays, Ready-To-Use Immunoassay
Bestellinformation
Shipping Condition: Wet ice
Spezifikation
Spezifikation
100 Assays
Fluorescent Detection
Tumor necrosis factor
Capture antibody coated beads, Detector antibody, Lot specific technical data sheet, Standard
<10 pg/mL
7124
Cell Culture Supernatant, Plasma, Serum
For research use only (RUO)