Learn More
Invitrogen™ Novex™ phosphoELISA™ GSK-3β (Total) ELISA Kit
Designed to detect and quantify the level of GSK-3β protein
Marke:  Invitrogen™ KHO0451
Weitere Details : Netto-Gewicht : 0.70000kg
Artikelnummer. 10571465
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beinhaltet: GSK-3β Antibody-Coated 96-Well Plate; GSK-3β Total Standard; Standard Diluent Buffer; GSK-3β Total Detection Antibody; anti-rabbit IgG-HRP (100X); HRP Diluent; Wash Buffer Concentrate (25X); Stabilized Chromogen, TMB; Stop Solution; Plate Covers; Detailed protocol with validation tests
Beschreibung
Beschreibung
The GSK-3β (Total) ELISA kit is intended for the detection of GSK-3β from human, rat and mouse cell lysates using 96-well plates and a microplate reader.
  • Easy to run Sandwich ELISA
  • More sensitive than Westerns
  • Convenient precoated, removable 8-well strips
  • Detect Low Expressing Proteins and Use Less Sample
  • Convenient Kit Format
  • Each ELISA kit is validated for sensitivity, specificity, precision, and lot-to-lot consistency
Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Drug Discovery and Development, Immunoassays, Kinase Antibodies and Immunoassays, Kinase Biology, Ready-To-Use Immunoassay, Target and Lead Identification and Validation
Bestellinformation
Shipping condition: Wet ice
Spezifikation
Spezifikation
1.6ng/mL
HRP
Serine/Threonine Kinases
2932
GSK-3β
10 μL
1.6 to 100ng/mL
Cell Extracts
Mouse, Rat
4 hr.
96 Assays
glycogen synthase kinase 3 beta