Learn More
Invitrogen HSP27 [pS82] Human ELISA Kit, 96 Tests The HSP27 Human ELISA Kit is intended to detect Shock Protein 27 (HSP27) phosphorylated at serine
Marke:  Invitrogen KHO0341
HSP27 [pS82] Human ELISA Kit, 96 Tests The HSP27 Human ELISA Kit is intended to detect Shock Protein 27 (HSP27) phosphorylated at serine
Weitere Details : Netto-Gewicht : 0.70000kg Transport : UN number : 3316 Chem class : 9 Pack group : III
Artikelnummer. 10583015