Learn More
Invitrogen™ Novex™ phosphoELISA™ JNK [pTpY183/185] Human ELISA Kit
Designed to detect and quantify the level of JNK protein phosphorylated at threonine residue 183 and tyrosine residue 185
Marke:  Invitrogen™ KHO0131
Weitere Details : Netto-Gewicht : 0.70000kg
Artikelnummer. 10533015
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »
Beinhaltet: JNK Antibody-Coated 96-Well Plate; JNK [pTpY183/185] Standard; Standard Diluent Buffer; JNK [pTpY183/185] Detection Antibody; anti-rabbit IgG-HRP (100X); HRP Diluent; Wash Buffer Concentrate (25X); Stabilized Chromogen, TMB; Stop Solution; Plate Covers; Detailed protocol with validation tests
Beschreibung
Beschreibung
The JNK [pTpY183/185] Human ELISA kit is intended for the detection of JNK [pTpY183/185] from human cell extracts using 96-well plates and a microplate reader.
  • Easy to run Sandwich ELISA
  • More sensitive than Westerns
  • Convenient precoated, removable 8-well strips
  • Detect Low Expressing Proteins and Use Less Sample
  • Convenient Kit Format
  • Each ELISA kit is validated for sensitivity, specificity, precision, and lot-to-lot consistency
Cancer Research, Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Clinical, Drug Discovery and Development, Immunoassays, Kinase Antibodies and Immunoassays, Kinase Biology, Oncology and Genetic Disease Research, Ready-To-Use Immunoassay, Target and Lead Identification and Validation
Bestellinformation
Shipping condition: Wet ice
Spezifikation
Spezifikation
<0.8 Units/mL
HRP
Serine/Threonine Kinases
5599
JNK
10 μL
1.6U/mL
Cell Extracts
Human
4 hr.
96 Assays
mitogen-activated protein kinase 8