Learn More
Invitrogen™ Novex™ phosphoELISA™ SMAD2 (Total) ELISA Kit
Designed to detect and quantify the level of SMAD2 protein
Marke:  Invitrogen™ KHO2021
Weitere Details : Netto-Gewicht : 0.47000kg
Artikelnummer. 10237354
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beinhaltet: SMAD2 Antibody-Coated 96-Well Plate; SMAD2 Total Standard; Standard Diluent Buffer; SMAD2 Total Detection Antibody; anti-rabbit IgG-HRP (100X); HRP Diluent; Wash Buffer Concentrate (25X); Stabilized Chromogen, TMB; Stop Solution; Plate Covers; Detailed protocol with validation tests
Beschreibung
Beschreibung
The SMAD2 (Total) ELISA kit is intended for the detection of SMAD2 from human cell lysates using 96-well plates and a microplate reader.
  • Easy to run Sandwich ELISA
  • More sensitive than Westerns
  • Convenient precoated, removable 8-well strips
  • Detect Low Expressing Proteins and Use Less Sample
  • Convenient Kit Format
  • Each ELISA kit is validated for sensitivity, specificity, precision, and lot-to-lot consistency
Cell Analysis, Drug Discovery and Development, Immunoassays, Kinase Antibodies and Immunoassays, Kinase Biology, Ready-To-Use Immunoassay, Target and Lead Identification and Validation
Bestellinformation
Shipping condition: Wet ice
Spezifikation
Spezifikation
< 0.067ng/mL
HRP
Transcription Factors
4087
SMAD2
10 μL
0.157 to 10ng/mL
Cell Extracts
Human
4 hr.
96 Assays
SMAD family member 2
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote
For Research Use Only. Not for use in diagnostic procedures.