Learn More
Invitrogen™ Novex™ phosphoELISA™ STAT5a [pY694] Human ELISA Kit
Designed to detect and quantify the levels of STAT5a phosphorylated at site tyrosine 694
Marke: Invitrogen™ KHO0761
Weitere Details : Netto-Gewicht : 0.10000kg
Beinhaltet: STAT5a Antibody-Coated 96-Well Plate; STAT5a Total Standard; Standard Diluent Buffer; STAT5a Total Detection Antibody; anti-rabbit IgG-HRP (100X); HRP Diluent; Wash Buffer Concentrate (25X); Stabilized Chromogen, TMB; Stop Solution; Plate Covers; Detailed protocol with validation tests
Beschreibung
Signal Transducer and Activator of Transcription 5 (STAT5) regulates gene transcription.
Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Drug Discovery and Development, Immunoassays, Kinase Antibodies and Immunoassays, Kinase Biology, Ready-To-Use Immunoassay, Target and Lead Identification and Validation
Bestellinformation
Shipping condition: Wet ice
Spezifikation
<1U/mL | |
HRP | |
Transcription Factors | |
6776 | |
STAT5a | |
10 μL |
1.6U/mL | |
Cell Extracts | |
Human | |
4 hr. | |
96 Assays | |
Signal transducer and activator of transcription 5A |
For Research Use Only. Not for use in diagnostic procedures.