missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NOXRED1 Polyclonal antibody specifically detects NOXRED1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Antigen | NOXRED1 |
| Anwendungen | Western Blot, Immunoprecipitation |
| Klassifikation | Polyclonal |
| Konjugat | Alexa Fluor 532 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | C14orf148, NADP-dependent oxidoreductase domain containing 1 |
| Wirtsspezies | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NOXRED1. |
| Reinigungsverfahren | Antigen and protein A Affinity-purified |
| Menge | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?