missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPBWR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 24625-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Interacts specifically with a number of opioid ligands. Receptor for neuropeptides B and W, which may be involved in neuroendocrine system regulation, food intake and the organization of other signals. Has a higher affinity for neuropeptide B.Spezifikation
NPBWR1 | |
Polyclonal | |
Unconjugated | |
NPBWR1 | |
G protein coupled receptor 7, GPR7, neuropeptides B/W receptor 1, NPBWR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2831 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P48145 | |
NPBWR1 | |
NPBWR1 Fusion Protein Ag16604 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |