missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human NTRK2 (TRKB), His Tag Recombinant Protein
Human NTRK2 (TRKB), His Tag Recombinant Protein
Marke: Thermo Scientific™ PV3616
Weitere Details : Netto-Gewicht : 0.05250kg
Beschreibung
NTRK2 (TRKB) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. NTRK2 is the receptor for brain-derived neurotrophic factor (BDNF). Together, NTRK2 and BDNF regulate both short-term synaptic functions and long-term potentiation of brain synapses.Spezifikation
NP_006171 | |
4915 | |
10 μg | |
Neurotrophic Receptor (NTRK) Kinase Family | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity | |
Dry Ice | |
NTRK2 | |
Baculovirus | |
Recombinant, Cytoplasmic | |
Receptor Tyrosine Kinases |
Kinase Assay | |
40.9 kDa | |
NTRK2 (TRKB) | |
aa 526-838 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 300 mM NaCl, 0.5 mM EDTA, 0.05% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
neurotrophic tyrosine kinase, receptor, type 2 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |