missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NUDT12 Recombinant Protein
Marke: Abnova™ H00083594-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH41099 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
76.56 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MSSVKRSLKQEIVTQFHCSAAEGDIAKLTGILSHSPSLLNETSENGWTALMYAARNGHPEIVQFLLEKGCDRSIVNKSRQTALDIAVFWGYKHIANLLATAKGGKKPWFLTNEVEECENYFSKTLLDRKSEKRNNSDWLLAKESHPATVFILFSDLNPLVTLGGNKESFQQPEVRLCQLNYTDIKDYLAQPEKITLIFLGVELEIKDKLLNYAGEVPREEEDGLVAWFALGIDPIAAEEFKQRHENCYFLHPPMPALLQLKEKEAGVVAQARSVLAWHSRYKFCPTCGNATKIEEGGYKRLCLKEDCPSLNGVHNTSYPRVDPVVIMQVIHPDGTKCLLGRQKRFPPGMFTCLAGFIEPGETIEDAVRREVEEESGVKVGHVQYVACQPWPMPSSLMIGCLALAVSTEIKVDKNEIEDARWFTREQVLDVLTKGKQQAFFVPPSRAIAHQLIKHWIRINPNL | |
DKFZp761I172 | |
NUDT12 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
83594 | |
NUDT12 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NUDT12 | |
Human | |
Recombinant | |
Solution |