Learn More
Nunc™ NucleoLink™ Strips
Ideal for hybridization assays due to the heat resistant resin, the unique breakable module strip design offers error-free flexibility, accurate reading and efficient washing.
Marke:  Thermo Scientific Nunc 248259
Weitere Details : Netto-Gewicht : 2.95000kg
Artikelnummer. 10040651
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beschreibung
Beschreibung
Ideal for hybridization assays due to the heat resistant resin, the unique design of Thermo Scientific™ Nunc™ NucleoLink™ Strips offer error-free flexibility; each well locks in the frame and maintains the same height to provide accurate reading and efficient washing.
Push the limits of your analysis and evaluate your performance with frames and strips that are ideal for binding nucleotides (e.g., DNA and RNA) and for hybridization assays.
  • Use in place of time-consuming methods such as gel electrophoresis and Southern blotting
  • Usable with any “ELISA-like” detection and measurement method
  • Compatible with standard microplate readers
  • Consists of 1 × 8 strips fitted into a 96-well format frame
  • V-shaped wells with flat, readable bottoms and thin walls (0.35mm)
  • Withstand temperatures from -20° to +120°C
  • Working volume: Strips 100μL